Anti-NRAP Rabbit Polyclonal Antibody
Catalog # 76757-448
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:nebulin-related anchoring protein
- Antigen Symbol:NRAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4892
- Antigen Synonyms:NRAP|N-RAP|nebulin-related anchoring protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NRAP (NP_001248392.1).
- Purification:Affinity purification.
- Cat. No.:76757-448
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human