Anti-SELENBP1 Rabbit Polyclonal Antibody
76757-958
: Prosci
- Antibody Type:Primary
- Antigen Name:selenium binding protein 1
- Antigen Symbol:SELENBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:8991
- Antigen Synonyms:hSP56|HEL-S-134P|LPSB|FLJ13813|SP56|SELENBP1|SBP56|hSBP
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 243-472 of human SELENBP1 (NP_003935.2).
- Purification:Affinity purification.
- Cat. No.:76757-958
- Packaging:Vial
Type: Primary
Antigen: SELENBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse