Anti-RRM1 Rabbit Polyclonal Antibody
Catalog # 76758-048
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:ribonucleotide reductase M1
- Antigen Symbol:RRM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:6240
- Antigen Synonyms:RIR1|RRM1|RR1|R1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 593-792 of human RRM1 (NP_001024.1).
- Purification:Affinity purification.
- Cat. No.:76758-048
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: RRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse