Anti-NUDT5 Rabbit Polyclonal Antibody
76755-554
: Prosci
- Antibody Type:Primary
- Antigen Name:nudix hydrolase 5
- Antigen Symbol:NUDT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:11164
- Antigen Synonyms:nuclear ATP-synthesis protein NUDIX5|YSA1|YSA1H|nudix (nucleoside diphosphate linked moiety X)-type motif 5|ADP-sugar pyrophosphatase|8-oxo-dGDP phosphatase|YSAH1|hNUDT5
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-219 of human NUDT5 (NP_054861.2).
- Purification:Affinity purification.
- Cat. No.:76755-554
- Packaging:Vial
Type: Primary
Antigen: NUDT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat