Anti-DDR2 Rabbit Polyclonal Antibody
Catalog # 76752-312
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:discoidin domain receptor tyrosine kinase 2
- Antigen Symbol:DDR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4921
- Antigen Synonyms:MIG20a|Hydroxyaryl-protein kinase|NTRKR3|DDR2|Discoidin domain receptor 2|Tyrosylprotein kinase|TYRO10|Tyrosine-protein kinase TYRO10|CD167b antigen
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 290-400 of human DDR2 (NP_001014796.1).
- Purification:Affinity purification.
- Cat. No.:76752-312
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat