Anti-UGT1A6 Rabbit Polyclonal Antibody
Catalog # 76764-876
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:UDP glucuronosyltransferase family 1 member A6
- Antigen Symbol:UGT1A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:54578
- Antigen Synonyms:UDP-glucuronosyltransferase 1-C|UDPGT 1-1|UGT-1C|UGT1.1|UDP-glucuronosyltransferase 1 family polypeptide A6s|UGT1F|UGT1-06|UGT1-01|HLUGP1|UDPGT 1-5|GNT1|UDP-glucuronosyltransferase 1A5|UGT1A3|UGT-1E|UDP-glucuronosyltransferase 1A3|UGT1E|UGT1A1|UDP-glucuronosyltransferase 1-5|Bilirubin-specific UDPGT isozyme 1|UGT1C|UDP-glucuronosyltransferase 1-6|UDP-glucuronosyltransferase 1-F|polypeptide A6|HLUGP|UGT1A6|UGT-1A|UDP-glucuronosyltransferase 1-1|hUG-BR1|UDP-glucuronosyltransferase 1-3|UGT-1F|UGT1.5|UDP-glucuronosyltransferase 1-E|UGT1.3|UGT1|UDP glucuronosyltransferase 1 family|UDP-glucuronosyltransferase 1A6|UDPGT 1-6|UGT1*6|UDP-glucuronosyltransferase 1-A|UDPGT 1-3|UGT1-05|phenol-metabolizing UDP-glucuronosyltransferase|UGT1-03|UGT1A|UDPGT|UDP glycosyltransferase 1 family|UGT1.6|UGT1A5S|UDP-glucuronosyltransferase 1A1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 65-270 of human UGT1A6 (NP_001063.2).
- Purification:Affinity purification.
- Cat. No.:76764-876
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: UGT1A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat