Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-Spike Protein Rabbit Recombinant Antibody [clone: 9C1A3]
  76761-110
 :  Prosci
Anti-Spike Protein Rabbit Recombinant Antibody [clone: 9C1A3]
  76761-110
 :  Prosci
 :  11-102
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Surface glycoprotein
  • Antigen Symbol:
    Spike Protein
  • Clonality:
    Recombinant
  • Clone:
    9C1A3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Virus
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    43740568
  • Antigen Synonyms:
    Spike glycoprotein
  • Modification:
    None
  • Storage Buffer:
    Supplied as a 0.2um filtered solution in PBS, pH 7.4.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    20
  • Immunogen:
    HEK293 derived SARS-COV-2 SPIKE RBD, amino acids Arg319-Phe541
    .
  • Purification:
    Affinity Purified, Endotoxin Level: < 1.0 EU/ug of the protein by LAL method.
  • Cat. No.:
    76761-110
  • Packaging:
    Vial

 

 

Type: Primary
Antigen: Spike protein
Clonality: Recombinant
Clone: 9C1A3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Virus