Anti-BMS-986004 Recombinant Antibody
Catalog # 76750-224
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Isotype:IgG1 kappa
- Size:0.1 mg
- Antigen Synonyms:GNR-011|AT-001|r84
- Modification:None
- Storage Buffer:PBS buffer pH 7.5 .
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Homo sapiens/VEGFA [Homo sapiens].
- Purification:>95%
- Cat. No.:76750-224
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity: