Anti-BMS-931699 Recombinant Antibody
Catalog # 76762-066
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Size:0.1 mg
- Antigen Synonyms:MT-112|BAY 2010112|BITE MT112
- Modification:None
- Storage Buffer:PBS buffer pH 7.5 .
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Chimeric,Humanized/CD3E/FOLH1 [Homo sapiens].
- Purification:>95%
- Cat. No.:76762-066
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: (scFv-heavy-kappa)-(scFv-heavy-lambda)
Reactivity: