Anti-SARS-CoV-2 Spike RBD Antibody Recombinant Antibody
Catalog # 76750-198
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Size:0.1 mg
- Gene ID:1043
- Antigen Synonyms:LDP-03|Campath-1H|CD52
- Modification:None
- Storage Buffer:PBS buffer pH 7.5 .
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8...730 °C for 1-2 weeks. Store at –20 …+730 °C for 12 months. Store at –80 …+730 °C for long term storage.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Humanized/CD52 [Homo sapiens].
- Purification:>95%
- Cat. No.:76750-198
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity: