To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ETS translocation varinat 4
- Antigen Symbol:ETV4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:2118
- Antigen Synonyms:PEA3|PEAS3|ETV4 Antibody: ETS variant 4|E1A-F|E1AF
- Modification:None
- Storage Buffer:ETV4 Antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:ETV4 antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human ETV4.
- Purification:ETV4 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:77133-114
- Packaging:Vial
Specifications
About this item
ETV4 antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type: Primary
Antigen: ETV4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat