Anti-DDDDK-Tag Antibody Rabbit Polyclonal Antibody
77128-972
: Prosci
- Antibody Type:Primary
- Antigen Name:DDDDK-tag antibody
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen Synonyms:flag|DDDDK|DDDDK-tag|flag-tag
- Modification:None
- Storage Buffer:DDDDK-tag antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:DDDDK-tag antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:DDDDK-tag antibody was raised against a synthetic peptide containing DYKDDDDK.
- Purification:DDDDK-tag antibody is affinity chromatography purified via peptide column.
- Cat. No.:77128-972
- Packaging:Vial
DDDDK-tag antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year.
Type: Primary
Antigen: DDDDK-tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tag