MIF Peptide
Catalog # 77127-818
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:4282
- Protein Synonyms:MMIF|Glycosylation-inhibiting factor|GLIF|Macrophage migration inhibitory factor|MIF|GIF
- Protein/Peptide Name:MIF
- Formulation:Lyophilized. MIF peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77127-818