AP-2 gamma Peptide
Catalog # 77125-320
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:7022
- Protein Synonyms:Activating enhancer-binding protein 2 gamma|hAP-2g|AP2-GAMMA|TFAP2G|ERF1|Transcription factor AP-2 gamma|AP2-gamma
- Protein/Peptide Name:TFAP2C
- Formulation:Lyophilized. AP-2 gamma peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77125-320