Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
GATA3 Peptide

GATA3 Peptide

Vial, 0.1 mg
Catalog # 77125-778
Supplier:   Prosci
Supplier #:  45-062P
GATA3 Peptide
Catalog # 77125-778
Supplier:  Prosci
CAS Number:  
GATA3 Peptide

Vial, 0.1 mg

CAS Number:  
Price
$301.99
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.1 mg
  • Storage Conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Cat. No.:
    77125-778
  • Shipping Temperature:
    RT
  • Protein/Peptide Type:
    Synthetic
  • Gene ID:
    2625
  • Formulation:
    Lyophilized. GATA3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Tested Applications:
    Blocking
  • Protein Synonyms:
    HDRS|HDR|Trans-acting T-cell-specific transcription factor GATA-3|GATA-binding factor 3
  • Protein/Peptide Name:
    GATA3
  • Conjugation:
    Unconjugated
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.