Human Recombinant sCD4 Protein (from CHO cells)
Catalog # 77126-104
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:10 µg
- Storage Conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological Activity:Determined by its ability to binds HeLa cells expressing cell-surface MHC-II molecules.
- Gene ID:920
- Protein Synonyms:OKT4(a-f)|Leu-3(a|b)|T-cell surface antigen T4/Leu-3|T-cell surface glycoprotein CD4|Cluster determinant 4|L3T4 (mouse)|Interleukin IL-16 receptor
- Protein/Peptide Name:CD4
- Purity:≥95% by SDS-PAGE gel and HPLC analyses
- Sequence:KKVVLGKKGD TVELTCTASQ KKSIQFHWKN SNQIKILGNQ GSFLTKGPSK LNDRADSRRS LWDQGNFPLI IKNLKIEDSD TYICEVEDQK EEVQLLVFGL TANSDTHLLQ GQSLTLTLES PPGSSPSVQC RSPRGKNIQG GKTLSVSQLE LQDSGTWTCT VLQNQKKVEF KIDIVVLAFQ KASSIVYKKE GEQVEFSFPL AFTVEKLTGS GELWWQAERA SSSKSWITFD LKNKEVSVKR VTQDPKLQMG KKLPLHLTLP QALPQYAGSG NLTLALEAKT GKLHQEVNLV VMRATQLQKN LTCEVWGPTS PKLMLSLKLE NKEAKVSKRE KAVWVLNPEA GMWQCLLSDS GQVLLESNIK VLPTWSTPVQ P
- Formulation:Lyophilized
- Shipping Temperature:RT
- Cat. No.:77126-104