Influenza A Recombinant Influenza A Strain A/Panama/2007/99 (H3N2) Antigen (from Embryonated chicken eggs)
Catalog # 77123-580
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Embryonated chicken eggs
- Species:Influenza A
- Size:0.05 mg
- Storage Conditions:Upon receipt, store at −20 °C. Avoid multiple freeze-thaw cycles.
- Protein/Peptide Name:Strain A/Panama/2007/99 (H3N2)
- Purity:>90% pure (SDS-PAGE). Ultracentrifugation using 10 - 40% sucrose gradient.
- Concentration:0.8 mg/ml (BCA method)
- Formulation:Liquid. 0.05 M Tris-HCl, pH 8.0 containing 0.1 M Sodium Chloride, 5 mM Ethylenediaminetetraacetic Acid, (EDTA)
- Shipping Temperature:20
- Tested Applications:EIA
- Cat. No.:77123-580