Influenza A Recombinant Influenza A Strain A/Kiev/301/94 (H3N2) Antigen (from Embryonated chicken eggs)
77128-818
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Embryonated chicken eggs
- Species:Influenza A
- Size:0.1 mg
- Storage Conditions:Upon receipt, store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/Peptide Name:Strain A/Kiev/301/94 (H3N2)
- Purity:>90% pure (SDS-PAGE). Ultracentrifugation using 10 - 40% sucrose gradient
- Concentration:1.7 mg/ml (BCA method)
- Formulation:Liquid. 0.05 M Tris-HCl, pH 8.0 containing 0.1 M Sodium Chloride, 5 mM EDTA
- Shipping Temperature:20
- Tested Applications:EIA
- Cat. No.:77128-818