Human Recombinant R-Spondin 3/RSPO3 (22-146) Protein (from HEK293 cells)
Catalog # 77125-180
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.05 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:84870
- Protein Synonyms:THSD2|PWTSR|CRISTIN1|RSPO3
- Protein/Peptide Name:RSPO3
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:15.9 kDa
- Sequence:Gln 22 - Val 146
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77125-180