Mouse Recombinant VEGF R1/Flt-1 Protein (from HEK293 cells)
77121-208
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:14254
- Protein Synonyms:FLT|FLT1|VEGFR1
- Protein/Peptide Name:Flt1
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:109.8 kDa
- Sequence:Tyr 23 - Glu 759
- Formulation:Lyophilized. 50 mM Tris, 100 mM Glycine, pH 7.5
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77121-208