HCoV-229E Recombinant S1 Protein (from HEK293 cells)
77119-798
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:HCoV-229E
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein Synonyms:Spike|Spike glycoprotein Subunit1|S glycoprotein Subunit1|S1 protein|Spike protein S1|COVID-19
- Protein/Peptide Name:HCoV-229E S1
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:58.8 kDa
- Sequence:Cys 16 - Ala 536
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77119-798