Rat Recombinant 4-1BB/TNFRSF9 Protein (from HEK293 cells)
Catalog # 77123-092
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein Synonyms:CD137|TNFRSF9|ILA|CDw137|4-1BB
- Protein/Peptide Name:4-1BB/TNFRSF9
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:19.5 kDa
- Sequence:Thr 24 - Gln 187
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77123-092