Anti-OSBPL5 Rabbit Polyclonal Antibody
Catalog # 76754-594
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Oxysterol binding protein-like 5
- Antigen Symbol:OSBPL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:114879
- Antigen Synonyms:oxysterol-binding protein-related protein 5|OSBP-related protein 5|oxysterol binding protein-like 5|FLJ31948|KIAA1534|OBPH1|OSBPL5|ORP5|oxysterol-binding protein homologue 1|DKFZp586H1719
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant protein of human OSBPL5.
- Purification:Affinity purification
- Cat. No.:76754-594
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: OSBPL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat