Anti-CAPRIN2 Rabbit Polyclonal Antibody
Catalog # 76761-514
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Caprin family member 2
- Antigen Symbol:CAPRIN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:65981
- Antigen Synonyms:RNA granule protein 14cytoplasmic activation/proliferation-associated protein 2|RNG14caprin-2|EEG1|EEG-1|C1q domain-containing protein 1|C1QDC1|gastric cancer multidrug resistance-associated protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant protein of human CAPRIN2.
- Purification:Affinity purification
- Cat. No.:76761-514
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CAPRIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat