Anti-VIRMA Rabbit Polyclonal Antibody
76762-980
: Prosci
- Antibody Type:Primary
- Antigen Name:Vir like m6A methyltransferase associated
- Antigen Symbol:VIRMA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:25962
- Antigen Synonyms:MSTP054|virilizer homolog|fSAP121|functional spliceosome-associated protein 121|protein virilizer homolog|KIAA1429
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1700 of human KIAA1429 (NP_056311.2).
- Purification:Affinity purification
- Cat. No.:76762-980
- Packaging:Vial
Type: Primary
Antigen: VIRMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey