Anti-ATPAF2 Rabbit Polyclonal Antibody
Catalog # 76749-840
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:ATP synthase mitochondrial F1 complex assembly factor 2
- Antigen Symbol:ATPAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:91647
- Antigen Synonyms:ATP12 homolog|MC5DN1|ATP12|ATP synthase mitochondrial F1 complex assembly factor 2|LP3663|ATP12p
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 41-200 of human ATPAF2 (NP_663729.1).
- Purification:Affinity purification
- Cat. No.:76749-840
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ATPAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse