Anti-DCTN5 Rabbit Polyclonal Antibody
Catalog # 76762-368
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Dynactin subunit 5
- Antigen Symbol:DCTN5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:84516
- Antigen Synonyms:dynactin subunit p25|dynactin subunit 5|dynactin 4|dynactin 5 (p25)
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DCTN5 (NP_115875.1).
- Purification:Affinity purification
- Cat. No.:76762-368
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DCTN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat