Anti-MMADHC Rabbit Polyclonal Antibody
Catalog # 76761-440
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria
- Antigen Symbol:MMADHC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:27249
- Antigen Synonyms:CL25022|cblD|C2orf25
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human MMADHC (NP_056517.1).
- Purification:Affinity purification
- Cat. No.:76761-440
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MMADHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat