Anti-TKFC Rabbit Polyclonal Antibody
Catalog # 76759-352
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Triokinase and FMN cyclase
- Antigen Symbol:TKFC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:26007
- Antigen Synonyms:dihydroxyacetone kinase 2 homolog|Dha kinase/FMN cyclase|DHA kinase|NET45|bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing)|triokinase/FMN cyclase|DAK|ATP-dependent dihydroxyacetone kinase|FAD-AMP lyase cyclic FMN forming|glycerone kinase|FAD-AMP lyase cyclizing|testis tissue sperm-binding protein Li 84P
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 226-575 of human TKFC (NP_056348.2).
- Purification:Affinity purification
- Cat. No.:76759-352
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TKFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat