Anti-RPP21 Rabbit Polyclonal Antibody
76754-864
: Prosci
- Antibody Type:Primary
- Antigen Name:Ribonuclease P/MRP subunit p21
- Antigen Symbol:RPP21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:79897
- Antigen Synonyms:RNaseP protein p21|C6orf135|CAT6ribonuclease P protein subunit p21|ribonucleoprotein V|ribonuclease P/MRP 21kDa subunit
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-154 of human RPP21 (NP_079115.1).
- Purification:Affinity purification
- Cat. No.:76754-864
- Packaging:Vial
Type: Primary
Antigen: RPP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse