Anti-SLC25A23 Rabbit Polyclonal Antibody
Catalog # 76754-838
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Solute carrier family 25 member 23
- Antigen Symbol:SLC25A23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:79085
- Antigen Synonyms:mitochondrial Ca2+-dependent solute carrier protein 2|APC2|SCaMC-3|mitochondrial ATP-Mg/Pi carrier protein 2|short calcium-binding mitochondrial carrier 3|small calcium-binding mitochondrial carrier protein 3|solute carrier family 25 (mitochondrial carrier|member 23|MCSC2|mitochondrial Ca(2+)-dependent solute carrier protein 2|calcium-binding mitochondrial carrier protein SCaMC-3|phosphate carrier)
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human SLC25A23 (NP_077008.2).
- Purification:Affinity purification
- Cat. No.:76754-838
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC25A23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat