Anti-VPS37A Rabbit Polyclonal Antibody
Catalog # 76752-710
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Vacuolar protein sorting 37 homolog A (S. cerevisiae)
- Antigen Symbol:VPS37A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:137492
- Antigen Synonyms:VPS37A|HCRP1|FLJ32642|FLJ42616|PQBP2|SPG53
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 224-397 of human VPS37A (NP_689628.2).
- Purification:Affinity purification
- Cat. No.:76752-710
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: VPS37A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat