Anti-KLHDC3 Rabbit Polyclonal Antibody
76750-956
: Prosci
- Antibody Type:Primary
- Antigen Name:Kelch domain containing 3
- Antigen Symbol:KLHDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:116138
- Antigen Synonyms:dJ20C7.3|testis intracellular mediator protein|PEAS|kelch domain-containing protein 3
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-382 of human KLHDC3 (NP_476502.1).
- Purification:Affinity purification
- Cat. No.:76750-956
- Packaging:Vial
Type: Primary
Antigen: KLHDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat