Anti-TLR2 Mouse Monoclonal Antibody [clone: [2B4A1]] (PE (Phycoerythrin))
Catalog # 77122-064
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:toll-like receptor 2
- Antigen Symbol:TLR2
- Clonality:Monoclonal
- Clone:[2B4A1]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:7097
- Antigen Synonyms:Toll-like Receptor 2
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping Temperature:20
- Cat. No.:77122-064
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: [2B4A1]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human