Anti-CD36 Mouse Monoclonal Antibody [clone: [SMf]] (FITC (Fluorescein Isothiocyanate))
Catalog # 77134-608
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Platelet glycoprotein 4
- Antigen Symbol:CD36
- Clonality:Monoclonal
- Clone:[SMf]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:lgM
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:948
- Antigen Synonyms:GPIV|PASIV|GPIIIb|OKM5-antigen
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Tonsil cells and peripheral blood mononuclear cells.
- Cat. No.:77134-608
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: [SMf]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: lgM
Reactivity: Human