Anti-MS4A1 Mouse Monoclonal Antibody [clone: [B-Ly1]] (PE (Phycoerythrin))
Catalog # 77122-278
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:B-lymphocyte antigen CD20
- Antigen Symbol:MS4A1
- Clonality:Monoclonal
- Clone:[B-Ly1]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Cat
- Size:100 tests
- Form:Liquid
- Gene ID:931
- Antigen Synonyms:MS4A1|Bp35|S7|B1|Leu-16
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping Temperature:20
- Cat. No.:77122-278
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: [B-Ly1]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Cat