Anti-Mouse Monoclonal Antibody [clone: [kit2c75]] (PE (Phycoerythrin))
Catalog # 77127-074
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Monoclonal
- Clone:[kit2c75]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Chicken
- Size:0.1 mg
- Form:Liquid
- Antigen Synonyms:CD117
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Immunogen:Sp/chkit4A4 cells (Sp2/0 cells transfected with chicken c-Kit cDNA).
- Cat. No.:77127-074
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: [kit2c75]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken