Anti-CD3D Mouse Monoclonal Antibody [clone: [CT-3]] (PE (Phycoerythrin))
Catalog # 77129-612
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CD3d molecule
- Antigen Symbol:CD3D
- Clonality:Monoclonal
- Clone:[CT-3]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Pigeon
- Size:0.1 mg
- Form:Liquid
- Gene ID:396518
- Antigen Synonyms:T3/TCR complex
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Immunogen:Chicken thymocytes and Ig- blood mononuclear cells.
- Cat. No.:77129-612
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD3D
Clonality: Monoclonal
Clone: [CT-3]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Pigeon