Anti-Mouse Monoclonal Antibody [clone: [BB6-10A10]] (PE (Phycoerythrin))
Catalog # 77127-032
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Monoclonal
- Clone:[BB6-10A10]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:lgM
- Reactivity:Pig
- Size:0.1 mg
- Form:Liquid
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Tested Applications:Blocking, Separation, Complement Mediated Cell Depletion
- Cat. No.:77127-032
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: [BB6-10A10]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: lgM
Reactivity: Pig