Anti-H2-K1 Mouse Monoclonal Antibody [clone: [16-3-22S]] (FITC (Fluorescein Isothiocyanate))
77123-814
: Prosci
- Antibody Type:Primary
- Antigen Name:histocompatibility 2, K1, K region
- Antigen Symbol:H2-K1
- Clonality:Monoclonal
- Clone:[16-3-22S]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:14972
- Antigen Synonyms:MHC Class I
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping Temperature:20
- Immunogen:C3H mouse splenocytes.
- Cat. No.:77123-814
- Packaging:Vial
Type: Primary
Antigen: H2-K1
Clonality: Monoclonal
Clone: [16-3-22S]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse