Anti-H2-Ea-ps Mouse Monoclonal Antibody [clone: [14-4-4S]] (FITC (Fluorescein Isothiocyanate))
Catalog # 77122-496
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:histocompatibility 2, class II antigen E alpha, pseudogene
- Antigen Symbol:H2-Ea-ps
- Clonality:Monoclonal
- Clone:[14-4-4S]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rat,Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:100504404
- Antigen Synonyms:MHC Class II
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping Temperature:20
- Immunogen:C3H mouse skin graft and splenocytes.
- Cat. No.:77122-496
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: H2-Ea-ps
Clonality: Monoclonal
Clone: [14-4-4S]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat