Anti-Klrb1c Mouse Monoclonal Antibody [clone: [PK136]] (PE (Phycoerythrin))
Catalog # 77130-114
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:killer cell lectin-like receptor subfamily B member 1C
- Antigen Symbol:Klrb1c
- Clonality:Monoclonal
- Clone:[PK136]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.1 mg
- Form:Liquid
- Gene ID:17059
- Antigen Synonyms:Ly-55|CD161b/CD161c
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Immunogen:CE mouse spleen enriched for NK-1+ cells and bone marrow cells.
- Cat. No.:77130-114
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Klrb1c
Clonality: Monoclonal
Clone: [PK136]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse