Anti-Cd38 Rat Monoclonal Antibody [clone: [NIMR-5]] (PE (Phycoerythrin))
Catalog # 77123-752
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CD38 antigen
- Antigen Symbol:Cd38
- Clonality:Monoclonal
- Clone:[NIMR-5]
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.1 mg
- Form:Liquid
- Gene ID:12494
- Antigen Synonyms:T10|cyclic ADP-ribose hydrolase|ADP-ribosyl cyclase
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Immunogen:BCL1 plasma membrane glycoproteins.
- Cat. No.:77123-752
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Cd38
Clonality: Monoclonal
Clone: [NIMR-5]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse