Anti-Snap23 Rabbit Monoclonal Antibody [clone: A6 ]
Catalog # 76750-622
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:synaptosomal-associated protein 23
- Antigen Symbol:Snap23
- Clonality:Monoclonal
- Clone:A6
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Gene ID:20619
- Antigen Synonyms:Sndt|23kDa|AA408749|Syndet|SNAP-23
- Modification:None
- Storage Buffer:PBS, pH 7.2, 0.02% NaN3.
- Storage Temperature:Store at 4 °C for up to 6 months. Store at −20 °C long term.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Synthetic peptide derived from mouse Snap23 (CRTKNFESGKNYKA).
- Purification:Protein A purified.
- Cat. No.:76750-622
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Snap23
Clonality: Monoclonal
Clone: A6
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse