Anti-NPPB Rabbit Monoclonal Antibody [clone: A10 ]
Catalog # 76749-114
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:natriuretic peptide B
- Antigen Symbol:NPPB
- Clonality:Monoclonal
- Clone:A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Gene ID:4879
- Antigen Synonyms:BNP
- Modification:None
- Storage Buffer:PBS, pH 7.2, 0.02% NaN3.
- Storage Temperature:Store at 4 °C for up to 6 months. Store at −20 °C long term.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Synthetic peptide derived from human NT-proBNP/C-terminus.
- Purification:Protein A purified.
- Cat. No.:76749-114
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NPPB
Clonality: Monoclonal
Clone: A10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human