Anti-neuron navigator 2 Goat Polyclonal Antibody
Catalog # 77130-436
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:neuron navigator 2
- Antigen Symbol:neuron navigator 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Size:0.1 mg
- Form:Liquid
- Gene ID:89797
- Antigen Synonyms:retinoic acid inducible gene in neuroblastoma 1|APC down-regulated 1|HELAD1|pore membrane and/or filament-interacting-like protein 2|POMFIL2|UNC53H2|NAV2|helicase|unc-53 homolog 2|neuron navigator 2|steerin-2|RAINB1|STEERIN2
- Modification:None
- Storage Buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage Temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping Temperature:20
- Immunogen:The immunogen for this antibody is: PELNCKGNGTAQS.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77130-436
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: neuron navigator 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: