Anti-MRG/MORF4L binding protein Goat Polyclonal Antibody
77125-876
: Prosci
- Antibody Type:Primary
- Antigen Name:MRG/MORF4L binding protein
- Antigen Symbol:MRG/MORF4L binding protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:55257
- Antigen Synonyms:OTTHUMP00000031512|FLJ10914|MRG15BP|URCC4|C20orf20|MRGBP|chromosome 20 open reading frame 20|MRG(MORF4-related gene)-binding protein|Eaf7|MRG-binding protein
- Modification:None
- Storage Buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage Temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping Temperature:20
- Immunogen:The immunogen for this antibody is: CIRDKFSQNIGRQVP.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77125-876
- Packaging:Vial
Type: Primary
Antigen: MRG/MORF4L binding protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human