Anti-SLC2A13 Rabbit Polyclonal Antibody
Catalog # 76764-438
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:solute carrier family 2 (facilitated glucose transporter), member 13
- Antigen Symbol:SLC2A13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:114134
- Antigen Synonyms:HMIT|MGC48624|SLC2A13
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 564-648 of human SLC2A13 (NP_443117.3).
- Purification:Affinity purification.
- Cat. No.:76764-438
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC2A13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat