Anti-GLDC Rabbit Polyclonal Antibody
Catalog # 76754-702
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Glycine dehydrogenase (decarboxylating), mitochondrial
- Antigen Symbol:GLDC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:2731
- Antigen Synonyms:GLDC|Glycine dehydrogenase (aminomethyl-transferring)|GCSP|Glycine decarboxylase|mitochondrial|Glycine dehydrogenase (decarboxylating)|Glycine cleavage system P protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 36-290 of human GLDC (NP_000161.2).
- Purification:Affinity purification.
- Cat. No.:76754-702
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: GLDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat