Anti-MAGI3 Rabbit Polyclonal Antibody
Catalog # 76753-528
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:membrane associated guanylate kinase, WW and PDZ domain containing 3
- Antigen Symbol:MAGI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:260425
- Antigen Synonyms:membrane-associated guanylate kinase|dJ730K3.2|MAGI-3|membrane-associated guanylate kinase inverted 3|WW and PDZ domain-containing protein 3|membrane-associated guanylate kinase-related 3
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human MAGI3 (NP_001136254.1).
- Purification:Affinity purification.
- Cat. No.:76753-528
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MAGI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse